di-Silberoxid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: di-Silver oxide, Disilver oxide, Silver oxide

11407.14 11407.22 11407.36
11407.14EA 175 CHF
11407.14 11407.22 11407.36
di-Silberoxid ≥99% (Metall-Basis)
di-Silberoxid

Purity: 99+% CAS No.: 20667-12-3 Molecular Formula: Ag2O Formula Weight: 231.74 Physical Form: Powder Density: 7.2 Appearance: Dark brown Odour: Odourless Melting Point: 230° dec. Merck Reference: 14,8521 UN No.: 1479 MDL No.: MFCD00003404


Polishing glass, Colouring glass yellow, catalyst, purifying drinking water, lab reagent

Formel: Ag₂O
MW: 231.74 g/mol
Melting Pt: 230 °C (Zers.)
Dichte: 7.2
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003404
CAS-Nummer: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1 II
Merck Index: 14,8521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR