VWR®, Laboratory Coat, Fluid-Impervious
Lieferant: VWR Collection
113-1117EA 125 CHF
113-1117
VWR®, Laboratory Coat, Fluid-Impervious
Einweg-Kleidung
Fluid-impervious lab coats are the cleanest and most durable within the entire VWR® Protection line of apparel.
  • Fluid-impervious material designed to repel water
  • Provides maximum protection
  • Excellent particulate barrier
  • Low-linting
Engineered to optimise barrier protection, cleanliness, and comfort, these garments are appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.
Lab coats feature snap front closure, knit cuffs, and three front pockets.
Zertifizierungen: Lab coats are rigorouslyMore Product Information