Bajonettfilter, X-plore®
Lieferant: Dräger
6738038 6738873 6738872 6738817 6738816 6738775 6738776 6738778 6738039 6738874
111-1046EA 3.1 CHF
111-1046 111-1056 111-1055 111-0770 111-1054 111-1049 111-1052 111-2347 DRAE6738039 DRAE6738874
Bajonettfilter, X-plore®
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
Sortiment an Filtern mit einem Dräger-spezifischen Bajonettanschluss für X-plore® 3300/3500 Halbmasken und X-plore® 5500 Vollmasken.
  • High level of user safety, great fit under face shields and visors
  • Robust gas filter with a blue ABS plastic housing (boiling point >65 °C)
  • White particle filter housing for easy identification of P3 filters
  • Option to upgrade a gas filter to a combination filter by simply adding a P1 or P2 pad or Dräger X-plore® Pure filter
  • Safe, quick and easy bayonet connection with clear guidance for positioning the filter securely
More Product Information