Lithiumcarbonat 99,998 Metals basis exCa, Puratronic® Ca_0,002

Lieferant: Thermo Fisher Scientific
Achtung

10734.18 10734.30 10734.A1
10734.18EA 222 CHF
10734.18 10734.30 10734.A1
Lithiumcarbonat 99,998 Metals basis exCa, Puratronic® Ca_0,002
Lithium carbonate

Purity: 99.998% CAS No.: 554-13-2 Molecular Formula: Li2CO3 Formula Weight: 73.90 Physical Form: Powder Density: 2.11 Appearance: White Odour: Odourless pH: 10-11.5 Melting Point: 723° Boiling Point: 1310° dec. Merck Reference: 14,5527 MDL No.: MFCD00011084

Formel: Li₂CO₃
MW: 73.89 g/mol
Siedepunkt: 1310 °C (Zers.)
Melting Pt: 723 °C
Dichte: 2.11
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011084
CAS-Nummer: 554-13-2
EINECS: 209-062-5
Merck Index: 14,5527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR