Indium ≥99,9995% (Metall-Basis), Perlen, Puratronic® ≤ 5 mm (0.2 in)

Lieferant: Thermo Fisher Scientific

10613.09 10613.18
10613.09EA 252 CHF
10613.09 10613.18
Indium ≥99,9995% (Metall-Basis), Perlen, Puratronic® ≤ 5 mm (0.2 in)
Indium

Purity: 99.9995% CAS No.: 7440-74-6 Odour: Odourless MDL No.: MFCD00134048

Formel: In
MW: 114.82 g/mol
Siedepunkt: 2000
Melting Pt: 156
Dichte: 7.3 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00134048
CAS-Nummer: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR