Titan 99,7 Metals basis, wire, Ø 0.25 mm (0.01 in)

Lieferant: Thermo Fisher Scientific
Achtung

10396.G6 10396.H2
10396.G6EA 104 CHF
10396.G6 10396.H2
Titan 99,7 Metals basis, wire, Ø 0.25 mm (0.01 in)
Titanium

Purity: 99.7% CAS No.: 7440-32-6 Physical Form: ≈0.22g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011264

Formel: Ti
MW: 47.88 g/mol
Siedepunkt: 3262
Melting Pt: 1660
Dichte: 4.51 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011264
CAS-Nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR