Bariumchlorid 0,1 mol/l (0,2 N) (20°C), Titripur®, Supelco®

Lieferant: Merck

1.60324.1003
1.60324.1003EA 120 CHF
1.60324.1003
Bariumchlorid 0,1 mol/l (0,2 N) (20°C), Titripur®, Supelco®
Barium chloride

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: BaCl₂
MW: 208.24 g/mol
Siedepunkt: 1560
Melting Pt: 963
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003445
CAS-Nummer: 10361-37-2
Merck Index: 12,00998

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR