HPLC columns, LiChroCART®, LiChrospher® 100 RP-8
Lieferant: Merck
1.50171.7088 1.50173.7087 1.50173.7088 1.50175.7087 1.50175.7088 1.50177.7087 1.50177.7088 1.50178.7087 1.50195.7087 1.50195.7088 1.50201.7087 1.50201.7088 1.50228.7087 1.50228.7088 1.50229.7087 1.50229.7088 1.50233.7087 1.50233.7088 1.50234.7087 1.50234.7088 1.50236.7087 1.50236.7088 1.50302.7087 1.50302.7088 1.51431.7088 1.51432.7088 1.51442.7088 1.51443.7087 1.51443.7088 1.51444.7087 1.51444.7088 1.51448.7088 1.50179.7088 1.50178.7088 1.50170.7088 1.50174.7088
1.50171.7088EA 619 CHF
1.50171.7088 1.50173.7087 1.50173.7088 1.50175.7087 1.50175.7088 1.50177.7087 1.50177.7088 1.50178.7087 1.50195.7087 1.50195.7088 1.50201.7087 1.50201.7088 1.50228.7087 1.50228.7088 1.50229.7087 1.50229.7088 1.50233.7087 1.50233.7088 1.50234.7087 1.50234.7088 1.50236.7087 1.50236.7088 1.50302.7087 1.50302.7088 1.51431.7088 1.51432.7088 1.51442.7088 1.51443.7087 1.51443.7088 1.51444.7087 1.51444.7088 1.51448.7088 1.50179.7088 1.50178.7088 1.50170.7088 1.50174.7088
HPLC columns, LiChroCART®, LiChrospher® 100 RP-8
Chromatographiesäulen
Stainless steel LiChroCART® cartridges are available with LiChrospher® sorbents. Ideal for separation of compounds under basic eluents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separateMore Product Information