Calciumchlorid Dihydrat, EMPROVE® ESSENTIAL BP, E509, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS
Achtung

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

1.42002.5000 1.42002.9029 1.42002.9525 1.42002.1000
1.42002.5000EA 458 CHF
1.42002.5000 1.42002.9029 1.42002.9525 1.42002.1000
Calciumchlorid Dihydrat, EMPROVE® ESSENTIAL BP, E509, SAFC®
Calciumchlorid Dihydrat

pH value: 4.5 - 8.5 (50 g/l, H₂O, 20 °C)
Solubility: 1280 g/l

Formel: CaCl₂·2H₂O
Siedepunkt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Dichte: 1.85 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149613
CAS-Nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR