Bathocuproinsulfonat Dinatriumsalz analytisches Reagens, Supelco®

Lieferant: Merck

Synonyms: Bathocuproinedisulphonic Acid Disodium Salt, Disodium Bathocuproinedisulphonate, Bathocuproine disulphonic acid disodium salt

M-Clarity™ Qualitätsstufe = MQ100

1.24518.0005
1.24518.0005EA 1100 CHF
1.24518.0005
Bathocuproinsulfonat Dinatriumsalz analytisches Reagens, Supelco®
Bathocuproinsulfonat Dinatriumsalz
Formel: C₂₆H₁₈N₂Na₂O₆S₂
MDL Number: MFCD00011679
CAS-Nummer: 52698-84-7
EINECS: 258-111-7

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) slightly soluble
Melting Point 300 °C
Molar Mass 564.55 g/mol
Bulk Density 190 kg/m3
pH 10 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR