Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel for atomic absorption spectroscopy (AAS), Supelco®
Lieferant: Merck
1.16755.1000
1.16755.1000EA 190.8 CHF
1.16755.1000
Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel for atomic absorption spectroscopy (AAS), Supelco®
Puffer Ionisationspuffer
10 g/L CsCl + 100 g/L La in dilute hydrochloric acid.