Hemacolor® Schnelles Färben von Blutausstrich solution_SC II staining reagent, Sigma-Aldrich®
Lieferant: Merck
1.11956.2500
1.11956.2500EA 157 CHF
1.11956.2500
Hemacolor® Schnelles Färben von Blutausstrich solution_SC II staining reagent, Sigma-Aldrich®
Hemacolor® Rapid staining of blood smear
Color reagent red (eosin solution)
Hemacolor® can be used in clinical cytology, e.g. for staining urine, sputum, FNAB, sperm, effusions and lavages, and in addition, for blood and bone marrow smears. The Hemacolor® staining kit is available with 100 ml or 500 ml bottles. It is developed and registered as CE / IVD product for applications in diagnostics. All Hemacolor solutions are additionally available as single 2.5 l PE-bottles. The package is sufficient for 1250 - 5000 applications. The buffer tablets acc. to Weise pH 7.2 are also available in larger pack sizes, thus, they can be useful for higher demands:
Hemacolor® solution 1, fixing solution
Hemacolor® solution 2, color reagent red
Hemacolor® solution 3, color reagent blue
IVDLogo