Tuerk solution Färbemittel für Hämatologie, Sigma-Aldrich®
Lieferant: Merck
Synonyms: Turk solution, Acetic acid gentian violet solution
M-Clarity™ quality level = MQ300
1.09277.0100
1.09277.0100EA 39.5 CHF
1.09277.0100
Tuerk solution Färbemittel für Hämatologie, Sigma-Aldrich®
Tuerk solution
Counting leukocytes in the counting chamber
The counting of leukocytes is an important part of routine hematological diagnosis. The basis of all counting methods is the dilution and preparation of a blood sample of known volume. The erythrocytes are hemolyzed by the acetic acid of Tuerk’s solution and the leukocytes are stained by the dye contained. The required cell type in a defined volume is counted and the number of cells per microliter of blood is then calculated. Tuerk's solution is an IVD product and CE registered.
IVDLogo