Zinkchlorid, EMSURE® Reag. Ph. Eur., ACS, ISO zur Analyse, Supelco®

Lieferant: Merck
Gefahr

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

M-Clarity™ quality level = MQ300

1.08816.0250 1.08816.1000 1.08816.9025
1.08816.0250EA 92.4 CHF
1.08816.0250 1.08816.1000 1.08816.9025
Zinkchlorid, EMSURE® Reag. Ph. Eur., ACS, ISO zur Analyse, Supelco®
Zinkchlorid
Formel: ZnCl₂
Siedepunkt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Dichte: 2.91 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011295
CAS-Nummer: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Spezifikation Testergebnisse

Solubility 4320 g/l (25 °C)
Melting Point 283 °C
Molar Mass 136.30 g/mol
Bulk Density 1400 - 1800 kg/m3
Boiling Point 732 °C (1013 hPa)
Vapor Pressure 1.33 hPa (428 °C)
Density 2.91 g/cm3 (25 °C)
pH 5 (100 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR