L(-)-Tryptophan, Millipore®

Lieferant: Merck

Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH

M-Clarity™ quality level = MQ100

1.08374.0010 1.08374.0100 1.08374.0500 1.08374.9010
1.08374.0010EA 67 CHF
1.08374.0010 1.08374.0100 1.08374.0500 1.08374.9010
L(-)-Tryptophan, Millipore®
L(-)-Tryptophan
Formel: C₁₁H₁₂N₂O₂
Melting Pt: 290 °C
Dichte: 1.34 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00064340
CAS-Nummer: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR