Pararosaniliniumchlorid, Certistain® für die Mikroskopie, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyms: Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride

1.07509.5000
1.07509.5000EA 0 CHF
1.07509.5000
Pararosaniliniumchlorid, Certistain® für die Mikroskopie, Sigma-Aldrich®
Pararosaniliniumchlorid

The PAS (periodic acid Schiff) reaction is one of the most frequently used chemical methods for histology. This Certistain® Pararosaniline (chloride) solid dye is used to prepare the Schiff’s reagent required for the PAS reaction, for the staining of mucopolysaccharides in histological tissue specimens. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.


The PAS (periodic acid Schiff) reaction is one of the most frequently used chemical methods for histology. This Certistain® Pararosaniline (chloride) solid dye is used to prepare the Schiff’s reagent required for the PAS reaction, for the staining of mucopolysaccharides in histological tissue specimens.
As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.

Formel: C₁₉H₁₈ClN₃
Melting Pt: 268 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001657
CAS-Nummer: 569-61-9
EINECS: 209-321-2
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR