Ammoniumdihydrogenphosphat 100 g/l in Wasser Phosphat-Matrixmodifikator für Graphitofen AAS, Supelco®

Lieferant: Merck

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

M-Clarity™ quality level = MQ100

1.07290.0050
1.07290.0050EA 130 CHF
1.07290.0050
Ammoniumdihydrogenphosphat 100 g/l in Wasser Phosphat-Matrixmodifikator für Graphitofen AAS, Supelco®
Ammoniumdihydrogenphosphat
Formel: NH₄H₂PO₄
MW: 115,03 g/mol
Siedepunkt: 100 °C
Dichte: 1.052 g/cm3 (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003396
CAS-Nummer: 7722-76-1

Order Now

Spezifikation Testergebnisse

Concentration ((NH₄)H₂PO₄) 100 ± 2 g/l

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR