di-Natriumhydrogenphosphat Dodecahydrat, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS

Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate

1.06573.5000 1.06573.9026 1.06573.9029 1.06573.9500
1.06573.5000EA 528 CHF
1.06573.5000 1.06573.9026 1.06573.9029 1.06573.9500
di-Natriumhydrogenphosphat Dodecahydrat, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®
di-Natriumhydrogenphosphat Dodecahydrat
Formel: Na₂HPO₄·12H₂O
Melting Pt: 35 °C
Dichte: 1.52 g/cm³ (20 °C)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00149181
CAS-Nummer: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR