Natriumiodid, Suprapur® Trace_analysis, Supelco®

Lieferant: Merck
Gefahr

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

MQ100

1.06519.0100
1.06519.0100EA 505 CHF
1.06519.0100
Natriumiodid, Suprapur® Trace_analysis, Supelco®
Natriumiodid
Formel: NaI
MW: 149.89 g/mol
Siedepunkt: 1300
Melting Pt: 662
Dichte: 3.67 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003532
CAS-Nummer: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Order Now

Spezifikation Testergebnisse

Solubility 1793 g/l (20 °C)
Melting Point 662 °C
Molar Mass 149.89 g/mol
Bulk Density 1500 - 2000 kg/m3
Boiling Point 1304 °C (1013 hPa)
Vapor Pressure 1.3 hPa (767 °C)
Density 3.67 g/cm3 (20 °C)
pH 6 - 9 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR