n-Heptan, EMPLURA®, Supelco®

Lieferant: Merck
Gefahr

Synonyms: high purity solvents

MQ200

1.04365.1000 1.04365.2500 1.04365.6025 1.04365.6190 1.04365.9950 1.04365.2511 1.04365.9011
1.04365.1000EA 93.6 CHF
1.04365.1000 1.04365.2500 1.04365.6025 1.04365.6190 1.04365.9950 1.04365.2511 1.04365.9011
n-Heptan, EMPLURA®, Supelco®
n-Heptan

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Formel: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Siedepunkt: 98.4
Melting Pt: –90.6 °C
Dichte: 0.6838 g/cm³ (20 °C)
Flash-Pt: –4 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009544
CAS-Nummer: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR