Gummi Arabicum, spray_dried, EMPROVE® ESSENTIAL BP, Ph. Eur. suitable for use as excipient, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS
Achtung

Synonyms: Acacia, Acacia gum

1.04228.9015
1.04228.9015EA 3550 CHF
1.04228.9015
Gummi Arabicum, spray_dried, EMPROVE® ESSENTIAL BP, Ph. Eur. suitable for use as excipient, SAFC®
Gummi Arabicum
Dichte: 1.35 g/cm³ (20 °C)
Flash-Pt: >250 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00081264
CAS-Nummer: 9000-01-5
EINECS: 232-519-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR