Guanidiniumchlorid, Millipore®

Lieferant: Merck
Achtung

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

1.04220.0100 1.04220.1000 1.04220.5000 1.04220.9025
1.04220.0100EA 89.9 CHF
1.04220.0100 1.04220.1000 1.04220.5000 1.04220.9025
Guanidiniumchlorid, Millipore®
Guanidiniumchlorid
Formel: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Dichte: 1.354 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013026
CAS-Nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR