Acetonitril, LiChrosolv® getestet für UHPLC-MS, Supelco®

Lieferant: Merck
Gefahr

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

1.03725.1002 1.03725.2002
1.03725.1002EA 254 CHF
1.03725.1002 1.03725.2002
Acetonitril, LiChrosolv® getestet für UHPLC-MS, Supelco®
Acetonitril

Highest purity solvents for UHPLC-MS analysis.


  • Suitability tested and specified for UHPLC-MS and UHPLC-UV
  • Specified quality in positive and negative ESI and APCI MS for lowest detection limits and confidence in analyses in all important MS modes - ESI / APCI (+) <2 ppb; ESI / APCI (-) <10 ppb)
  • Lowest impurity profile: For interference-free baselines
  • Lowest levels of trace metal impurities: For minimised metal ion adduct formation (up to <5 ppb)
  • Lowest level of polyethylene glycol (PEG) impurities in our entire UHPLC-MS solvent line up to give you confidence in your results (PEG S/N signal-to-noise ratio <50)


Microfiltration through 0,2 µm filter: Prolonged lifetime of filters and mechanical parts in HPLC systems and reduced risk of column clogging.
Packed in borosilicate glass bottles: Minimises contamination with metal ions.
The borosilicate bottle with GL 45 neck finish can be directly connected to instruments.

Formel: H₃CCN
MW: 41.05 g/mol
Siedepunkt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Dichte: 0.782 g/cm³ (20 °C)
Flash-Pt: 2 °C (geschlossener Tiegel)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001878
CAS-Nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Al (Aluminum) ≤ 5 ppb
Ca (Calcium) ≤ 5 ppb
Evaporation residue ≤ 0.7 mg/L
Fe (Iron) ≤ 5 ppb
Fluorescence at 254 nm ≤ 0.3 ppb
Fluorescence at 365 nm ≤ 0.3 ppb
ldentity (IR-spectrum) PASSES
K (Potassium) ≤ 5 ppb
Mg (Magnesium) ≤ 5 ppb
Na (Sodium) ≤ 25 ppb
Polyethyleneglycol (PEG) Signal-te-noise ratio ≤ 50
Purity (GC) ~ 99.9 %
Suitable for LC/MS; lntensity of background mass signals based on reserpine (APCI/ESI negative) ≤ 10 ppb
Suitable for LC/MS; lntensity of background mass signals based on reserpine (APCI/ESI positive) ≤ 2 ppb
Titrable acid ≤ 0.001 meq/g
Titrable base ≤ 0.0002 meq/g
UHPLC gradient (at 210nm) ≤ 1.0 mAU
UHPLC gradient (at 254nm) ≤ 0.2 mAU
Water (H2O) ≤ 0.01 %
Filtered by 0.2um filter


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR