1,4-Dioxan stabilisiert, EMPLURA®, Supelco®

Lieferant: Merck
Gefahr

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 25 ppm
M-Clarity™ quality level = MQ200

1.03115.1000 1.03115.2500 1.03115.6025 1.03115.9191
1.03115.1000EA 167 CHF
1.03115.1000 1.03115.2500 1.03115.6025 1.03115.9191
1,4-Dioxan stabilisiert, EMPLURA®, Supelco®
1,4-Dioxan
Formel: C₄H₈O₂
Siedepunkt: 101.1 °C (1013 hPa)
Melting Pt: 11.8 °C
Dichte: 1.03 g/cm³ (20 °C)
Flash-Pt: 12 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006571
CAS-Nummer: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3,II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR