Chloramin T (Natriumsalz) Trihydrat, reinst, Supelco®

Lieferant: Merck
Gefahr

Synonyms: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

M-Clarity™ quality level = MQ200

1.02424.1000 1.02424.5000 1.02424.9050
1.02424.1000EA 623 CHF
1.02424.1000 1.02424.5000 1.02424.9050
Chloramin T (Natriumsalz) Trihydrat, reinst, Supelco®
Chloramin T (Natriumsalz) Trihydrat
Formel: CH₃C₆H₄SO₂NClNa·3H₂O
Melting Pt: 167…170 °C
Dichte: 1.43 g/cm³ (20 °C)
Flash-Pt: 192 °C (closed cup)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00149066
CAS-Nummer: 7080-50-4
EINECS: 204-854-7
UN: 2928
ADR: 6.1 (8),II
Merck Index: 13,02085

Order Now

Spezifikation Testergebnisse

Assay (iodometric) 98.0 - 103.0 %
Identity (IR-spectrum) passes test
Appearance of solution passes test
pH-value (5 %; water) 8.0 - 10.0
Matter insoluble in ethanol ≤ 2 %
ortho compounds passes test
Residual solvents (ICH Q3C) excluded by manufactoring process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR