Calciumhydrogenphosphat, anhydrous, powder_extra_fine, EMPROVE® EXPERT FCC, BP, E341, Ph. Eur., USP excipient_suitable, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS

Synonyms: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic

1.02144.9026 1.02144.9025 1.02144.1000
1.02144.9026EA 0 CHF
1.02144.9026 1.02144.9025 1.02144.1000
Calciumhydrogenphosphat, anhydrous, powder_extra_fine, EMPROVE® EXPERT FCC, BP, E341, Ph. Eur., USP excipient_suitable, SAFC®
Calciumhydrogenphosphat
Formel: CaHPO₄
MW: 136.06 g/mol
Melting Pt: 370
Dichte: 2.89 g/cm³ (20 °C)
MDL Number: MFCD00010909
CAS-Nummer: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR