Salzsäure 30%, ultrapure Trace_analysis, Supelco®

Lieferant: Merck
Gefahr

Synonyms: Hydrochloric acid solution, Muriatic acid

MQ100

1.01514.0250 1.01514.0500 1.01514.1000
1.01514.0250EA 1140 CHF
1.01514.0250 1.01514.0500 1.01514.1000
Salzsäure 30%, ultrapure Trace_analysis, Supelco®
Salzsäure
Formel: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Dichte: 1.18 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Spezifikation Testergebnisse

Assay (acidimetric) ≥ 30.0 %
Ag (Silver) ≤ 100 ppt
Al (Aluminium) ≤ 100 ppt
As (Arsenic) ≤ 100 ppt
Au (Gold) ≤ 100 ppt
B (Boron) ≤ 100 ppt
Ba (Barium) ≤ 50 ppt
Be (Beryllium) ≤ 50.0 ppt
Bi (Bismuth) ≤ 50.0 ppt
Ca (Calcium) ≤ 500 ppt
Cd (Cadmium) ≤ 50.0 ppt
Ce (Cerium) ≤ 20.00 ppt
Co (Cobalt) ≤ 100.0 ppt
Cr (Chromium) ≤ 50 ppt
Cu (Copper) ≤ 100 ppt
Dy (Dysprosium) ≤ 20.00 ppt
Er (Erbium) ≤ 20.00 ppt
Eu (Europium) ≤ 20.00 ppt
Fe (Iron) ≤ 500 ppt
Ga (Gallium) ≤ 50.0 ppt
Gd (Gadolinium) ≤ 20 ppt
Hf(Hafnium) ≤ 10.00 ppt
Hg (Mercury) ≤ 50 ppt
Ho (Holmium) ≤ 20.00 ppt
In (Indium) ≤ 50.0 ppt
K (Potassium) ≤ 500 ppt
La (Lanthanum) ≤ 20.0 ppt
Li (Lithium) ≤ 50.0 ppt
Lu (Lutetium) ≤ 20.00 ppt
Mg (Magnesium) ≤ 100 ppt
Mn (Manganese) ≤ 50 ppt
Mo (Molybdenum) ≤ 50.0 ppt
Na (Sodium) ≤ 200 ppt
Nb (Niobium) ≤ 20.0 ppt
Nd (Neodymium) ≤ 20.0 ppt
Ni (Nickel) ≤ 200 ppt
Pb (Lead) ≤ 50.0 ppt
Pd (Palladium) ≤ 100 ppt
Pr (Praseodym) ≤ 20.00 ppt
Pt (Platinum) ≤ 50 ppt
Rb (Rubidium) ≤ 20.0 ppt
Re (Rhenium) ≤ 10.00 ppt
Ru (Ruthenium) ≤ 20.0 ppt
Sb (Antimony) ≤ 100 ppt
Sc (Scandium) ≤ 20 ppt
Se (Selenium) ≤ 200 ppt
Si (Silicon) ≤ 1000 ppt
Sm (Samarium) ≤ 20.00 ppt
Sn (Tin) ≤ 200 ppt
Sr (Strontium) ≤ 50.0 ppt
Ta (Tantalum) ≤ 100.00 ppt
Tb (Terbium) ≤ 20.00 ppt
Te (Tellur) ≤ 100 ppt
Th (Thorium) ≤ 1.00 ppt
Ti (Titanium) ≤ 50 ppt
Tl (Thallium) ≤ 50.0 ppt
Tm (Thulium) ≤ 20.00 ppt
U (Uranium) ≤ 1.00 ppt
V (Vanadium) ≤ 50 ppt
W (Wolfram) ≤ 100 ppt
Y (Yttrium) ≤ 10.0 ppt
Yb (Ytterbium) ≤ 20.0 ppt
Zn (Zinc) ≤ 200 ppt
Zr (Zirconium) ≤ 20.0 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR