Methylorange 0,1% aqueous solution, Supelco®

Lieferant: Merck

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

MQ200

1.01323.0250 1.01323.1000
1.01323.0250EA 112 CHF
1.01323.0250 1.01323.1000
Methylorange 0,1% aqueous solution, Supelco®
Methylorange
Formel: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Siedepunkt: 90
Flash-Pt: 38
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007502
CAS-Nummer: 547-58-0
EINECS: 208-925-3
Merck Index: 13,06127

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Density 1.00 g/cm3 (20 °C)
pH 6 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR