Titangelb, Supelco®

Lieferant: Merck

Synonyms: Disodium 2,2'-(1-triazene-1,3-diyldi-p-phenylene)bis[6-methylbenzothiazole-7-sulphonate], Direct Yellow 9, Clayton Yellow, Thiazole Yellow G

M-Clarity™ Qualitätsstufe = MQ200

1.01307.0025
1.01307.0025EA 136 CHF
1.01307.0025
Titangelb, Supelco®
Titangelb
Formel: C₂₈H₁₉N₅Na₂O₆S₄
MW: 695.73 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013091
CAS-Nummer: 1829-00-1
EINECS: 217-377-4
Merck Index: 12,09445

Order Now

Spezifikation Testergebnisse

Solubility 29 g/l (24 °C)
Melting Point 300 °C (decomposition)
Molar Mass 695.7 g/mol
Bulk Density 310 kg/m3
pH 5.1 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR