di-Ammoniumoxalat Monohydrat, Supelco®

Lieferant: Merck
Achtung

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

M-Clarity™ Qualitätsstufe = MQ200

1.01190.1000 1.01190.9050
1.01190.1000EA 191 CHF
1.01190.1000 1.01190.9050
di-Ammoniumoxalat Monohydrat, Supelco®
di-Ammoniumoxalat Monohydrat
Formel: (NH₄)₂C₂O₄·H₂O
MW: 142.11 g/mol
Siedepunkt: 70 °C (1013 hPa)
Melting Pt: 70 °C
Dichte: ∼1.5 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149694
CAS-Nummer: 6009-70-7
EINECS: 214-202-3
UN: 3288
ADR: 6.1 III
Merck Index: 13,00541

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR