Ammoniumdihydrogenphosphat, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

Lieferant: Merck

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

MQ300

1.01126.0500 1.01126.9050 1.01126.5000
1.01126.0500EA 36.18 CHF
1.01126.0500 1.01126.9050 1.01126.5000
Ammoniumdihydrogenphosphat, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
Ammoniumdihydrogenphosphat
Formel: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190
Dichte: 1.8 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003396
CAS-Nummer: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547

Order Now

Spezifikation Testergebnisse

Solubility 368 g/l (20 °C)
Melting Point 190 °C
Molar Mass 115.02 g/mol
Bulk Density 800 - 1100 kg/m3
Vapor Pressure 0.066 hPa (125 °C)
Density 1.80 g/cm3 (20 °C)
pH 3.8 - 4.4 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR