Aluminiumchlorid Hexahydrat, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra_pure suitable for use as excipient, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS
Gefahr

Synonyms: Aluminum chloride hexahydrate

1.01084.9026
1.01084.9026EA 0 CHF
1.01084.9026
Aluminiumchlorid Hexahydrat, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra_pure suitable for use as excipient, SAFC®
Aluminiumchlorid Hexahydrat
Formel: AlCl₃·6H₂O
MW: 241.43 g/mol
Melting Pt: ∼100 °C
Dichte: 2.39 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149134
CAS-Nummer: 7784-13-6
EINECS: 231-208-1
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR