1,1,1-Trichlor-2-methyl-2-propanol Hemihydrat BP, Ph. Eur., JP, NF, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS
Achtung

Synonyms: 1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate, Chlorbutanol hemihydrate, Chlorobutanol hemihydrate, Chlorobutanol hydrous

1.01005.1000
1.01005.1000EA 549 CHF
1.01005.1000
1,1,1-Trichlor-2-methyl-2-propanol Hemihydrat BP, Ph. Eur., JP, NF, SAFC®
1,1,1-Trichlor-2-methyl-2-propanol Hemihydrat
Formel: C₈H₁₆Cl₆O₃
Siedepunkt: 167 °C (1013 hPa)
Melting Pt: 78 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02179352
CAS-Nummer: 6001-64-5
EINECS: 200-317-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR