1-Octanol, EMPLURA®, Supelco®

Lieferant: Merck
Achtung

Synonyms: n-Octanol

M-Clarity™ quality level = MQ200

1.00991.1000 1.00991.6025
1.00991.1000EA 151 CHF
1.00991.1000 1.00991.6025
1-Octanol, EMPLURA®, Supelco®
1-Octanol
Formel: H₃C(CH₂)₆CH₂OH
Siedepunkt: 195 °C (1013 hPa)
Melting Pt: –16 °C
Dichte: 0.8246 g/cm³ (20 °C)
Flash-Pt: 81 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002988
CAS-Nummer: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR