Cetylalkohol, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, NF, reinst suitable for use as excipient, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS

Synonyms: Cetyl alcohol, Palmityl alcohol

1.00989.9024
1.00989.9024EA 1120 CHF
1.00989.9024
Cetylalkohol, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, NF, reinst suitable for use as excipient, SAFC®
1-Hexadecanol
Formel: H₃C(CH₂)₁₄CH₂OH
Siedepunkt: 305…330 °C (1013 hPa)
Melting Pt: 46…52 °C
Dichte: 0.81 g/cm³ (20 °C)
Flash-Pt: >110 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004760
CAS-Nummer: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR