Ethanol absolut ≥99,9% (durch GC), EMSURE® Reag. Ph. Eur., ACS, ISO zur Analyse, Supelco®

Lieferant: Merck
Gefahr

Synonyms: Ethyl alcohol

M-Clarity™ quality level = MQ300

1.00983.1000 1.00983.1011 1.00983.2500 1.00983.2511 1.00983.5000 1.00983.6010 1.00983.6025 1.00983.9180 1.00983.9950 1.00983.6190 1.00983.9025 1.00983.1000_T 1.00983.1011_T 1.00983.2500_T 1.00983.2511_T 1.00983.5000_T 1.00983.6025_T 1.00983.6010_T
1.00983.1000EA 83.1 CHF
1.00983.1000 1.00983.1011 1.00983.2500 1.00983.2511 1.00983.5000 1.00983.6010 1.00983.6025 1.00983.9180 1.00983.9950 1.00983.6190 1.00983.9025 MERC1.00983.1000_T MERC1.00983.1011_T MERC1.00983.2500_T MERC1.00983.2511_T MERC1.00983.5000_T MERC1.00983.6025_T MERC1.00983.6010_T
Ethanol absolut ≥99,9% (durch GC), EMSURE® Reag. Ph. Eur., ACS, ISO zur Analyse, Supelco®
Ethanol

Bio-Based Ethanol
Instead of synthetic ethanol, we use bioethanol produced from grain or sugar cane. High quality, affordability, and ready availability make our bioethanol an obvious choice for a sustainable future.


  • Produced from grain or sugar cane, a renewable source
  • Less toxic than synthetic ethanol (no toxic by-products)
  • Reliable availability
  • Production method is safer for the environment

Formel: C₂H₅OH
Siedepunkt: 78.3 °C (1013 hPa)
Melting Pt: –117 °C
Dichte: 0.7895 g/cm³ (20 °C)
Flash-Pt: 12 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003568
CAS-Nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Spezifikation Testergebnisse

Purity (GC) ≥ 99.9 %
Identity (IR) conforms
Appearance conforms
Color ≤ 10 Hazen
Solubility in water conforms
Acidity or alkalinity ≤ 30 ppm
Titrable acid ≤ 0.0002 meq/g
Titrable base ≤ 0.0002 meq/g
Density (d 20 °C/20 °C) 0.790 - 0.793
UV absorption conforms
Aldehydes (as Acetaldehyd) ≤ 0.001 %
Fusel oils conforms
Substances reducing potassium permanganate (as O) ≤ 0.0002 %
Substances reducing permanganate (ACS) conforms
Carbonyl compounds (as CO) ≤ 0.003 %
Readily carbonizable substances conforms
Acetone, Isopropyl Alcohol (ACS) conforms
Acetone (GC) ≤ 0.001 %
Ethylmethylketone (GC) ≤ 0.02 %
Isoamyl alcohol (GC) ≤ 0.05 %
2-Propanol (GC) ≤ 0.01 %
Higher alcohols (GC) ≤ 0.01 %
Volatile impurities (GC) (Acetaldehyde and Acetal) ≤ 10 ppm
Volatile impurities (GC) (Benzene) ≤ 2 ppm
Volatile impurities (GC) (Methanol) ≤ 100 ppm
Volatile impurities (GC) (Total of other impurities) ≤ 300 ppm
Volatile impurities (GC) (disregard limit) ≤ 9 ppm
Chloride (Cl) ≤ 0.3 ppm
Nitrate (NO₃) ≤ 0.3 ppm
Phosphate (PO₄) ≤ 0.3 ppm
Sulfate (SO₄) ≤ 0.3 ppm
Ag (Silver) ≤ 0.000002 %
Al (Aluminium) ≤ 0.00005 %
As (Arsenic) ≤ 0.000002 %
Au (Gold) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Be (Beryllium) ≤ 0.000002 %
Bi (Bismuth) ≤ 0.000002 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Ga (Gallium) ≤ 0.000002 %
In (Indium) ≤ 0.000002 %
Li (Lithium) ≤ 0.000002 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Mo (Molybdenum) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Pt (Platinum) ≤ 0.000002 %
Sb (Antimony) ≤ 0.000002 %
Sn (Tin) ≤ 0.00001 %
Ti (Titanium) ≤ 0.000002 %
Tl (Thallium) ≤ 0.000002 %
V (Vanadium) ≤ 0.000002 %
Zn (Zinc) ≤ 0.00001 %
Zr (Zirconium) ≤ 0.000002 %
Evaporation residue ≤ 0.0005 %
Water ≤ 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR