Ethylacetat, EMPROVE® ESSENTIAL NF, Ph. Eur., reinst suitable for use as excipient, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS
Gefahr

Synonyms: Acetic acid ethyl ester

1.00864.1000 1.00864.2500 1.00864.6010 1.00864.6025 1.00864.9181
1.00864.1000EA 109 CHF
1.00864.1000 1.00864.2500 1.00864.6010 1.00864.6025 1.00864.9181
Ethylacetat, EMPROVE® ESSENTIAL NF, Ph. Eur., reinst suitable for use as excipient, SAFC®
Ethylacetat
Formel: CH₃COOC₂H₅
Siedepunkt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Dichte: 0.902 g/cm³ (20 °C)
Flash-Pt: –4 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR