Ameisensäure 98 - 100% 98-100% (durch Alkalimetrie), EMPROVE® ESSENTIAL DAC, reinst suitable for use as excipient, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS
Gefahr

Synonyms: Aminic Acid, Methanoic acid

1.00263.1000 1.00263.2500 1.00263.9025
1.00263.1000EA 161 CHF
1.00263.1000 1.00263.2500 1.00263.9025
Ameisensäure 98 - 100% 98-100% (durch Alkalimetrie), EMPROVE® ESSENTIAL DAC, reinst suitable for use as excipient, SAFC®
Ameisensäure

Elemental impurity specifications have been set considering ICH Q3D (guideline for elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Zertifizierungen: Corresponds to DAC.

Formel: HCO₂H
MW: 46.03 g/mol
Melting Pt: 4 °C
Dichte: 1.22
Flash-Pt: 48
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003297
CAS-Nummer: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay (Alkalimetric) ≥98.0%
Identity (IR-Spectrum) Passes Test
Density (d 20/20) 1.217 - 1.223
Chromatographic Purity (Acetic acid) ≤0.5%
Chromatographic Purity (Other Impurity) ≤0.1%
Chromatographic Purity (Sum of All Other Impurities) ≤0.3%
Chloride (Cl) ≤0.001%
Sulfate (SO₄) ≤0.004%
Sulfite (SO₃) ≤0.002%
Heavy Metals (as Pb) ≤0.0005%
As (Arsenic) ≤0.0003%
Cu (Copper) ≤0.002%
Pb (Lead) ≤0.001%
Zn (Zinc) ≤0.002%
Aldehydes Passes Test
Formaldehyde (HCHO) ≤0.1%
Oxalic Acid (C₂H₂O₄) ≤0.01%
Residual Solvents (ICHQ3C) of Class 2 (Methanol) ≤0.3%
Evaporation Residue ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR