Aluminon, Supelco®

Lieferant: Merck

Synonyms: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt

M-Clarity™ quality level = MQ100

1.00128.0010
1.00128.0010EA 957 CHF
1.00128.0010
Aluminon, Supelco®
Aluminon
Formel: C₂₂H₂₃N₃O₉
MW: 473,44 g/mol
MDL Number: MFCD00040925
CAS-Nummer: 569-58-4
EINECS: 209-319-1

Order Now

Spezifikation Testergebnisse

Solubility 800 g/l (20 °C)
Melting Point 220 - 225 °C decomposes
Molar Mass 473.44 g/mol
Bulk Density 400 kg/m3
pH 5.3 (10 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR