Aceton, LiChrosolv® für die HPLC, Supelco®

Lieferant: Merck
Gefahr

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

M-Clarity™ quality level = MQ100

1.00020.1000 1.00020.2500 1.00020.4000 1.00020.9030 1.00020.9010
1.00020.1000EA 48.26 CHF
1.00020.1000 1.00020.2500 1.00020.4000 1.00020.9030 1.00020.9010
Aceton, LiChrosolv® für die HPLC, Supelco®
Aceton

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Formel: CH₃COCH₃
Siedepunkt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Dichte: 0.792 g/cm³ (20 °C)
Flash-Pt: <–20 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point -95.4 °C
Molar Mass 58.08 g/mol
Boiling Point 56.2 °C (1013 hPa)
Vapor Pressure 233 hPa (20 °C)
Flash Point
Refractive Index 1.35868 (20 °C)
Explosion Limit 2.6 - 12.8 %(V)
Density 0.79 g/cm3 (20 °C)
pH 5 - 6 (395 g/l, H2O, 20 °C)
Ignition Point 465 °C DIN 51794
Viscosity kinematic
Synonyms Dimethyl ketone, Propanone, 2-Propanone
Hill Formula C3H6O
Chemical Formula CH3COCH3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR