Dimethyldichlorsilan solution_SC ~5% toluene, LiChropur™ derivatisation_GC, Supelco®

Lieferant: Merck

Synonyms: Dichloro(dimethyl)silane, Dichlorodimethylsilane

08471-500ML
08471-500MLEA 147 CHF
08471-500ML
Dimethyldichlorsilan solution_SC ~5% toluene, LiChropur™ derivatisation_GC, Supelco®
Dimethyldichlorsilan

Sylon CT™ was used as derivatizing agent, during an experiment to develop sensitive GC method for the identification and determination of long-chain primary alkyl amines.† It was also used for treating Erlenmeyer flasks to prevent mycelium from sticking to the glassware during development of techniques in the identification and partial characterization the chaxamycin biosynthesis gene cluster of S. leeuwenhoekii.


  • Suitable for deactivating glassware


Treat the glass tubing that you use for columns with Sylon-CT solution (5% dimethyldichlorosilane in toluene). It deactivates the tubing for use up to 350 to 400 °C, far exceeding the capabilities of other deactivating agents used at room temperature. It also deactivates transfer lines and glass or glass-lined injection port liners. Instructions included.

Formel: C₂H₆Cl₂Si
MW: 129.06 g/mol
Siedepunkt: 70
Melting Pt: -76
Flash-Pt: -16
MDL Number: MFCD00000491
CAS-Nummer: 75-78-5
UN: 2920
ADR: 3(8) I

Order Now

Spezifikation Testergebnisse

Appearance (Color) Colorless to Almost Colorless
Appearance (Form) Liquid
Argentometr. Titration 4.5 - 5.5 %
Refractive Index N20/D 1.491 - 1.495


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR