Glycin (H-Gly-OH) 98,5-101,5 USP, multi-compendial, J.T.Baker®

Lieferant: Avantor

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

0582-01EA 190 CHF
0582-01 0582-05 0582-07 0582-09
Glycin (H-Gly-OH) 98,5-101,5 USP, multi-compendial, J.T.Baker®
Glycin (H-Gly-OH)
Formel: NH₂CH₂COOH
MW: 75.07 g/mol
Siedepunkt: 181
Melting Pt: 83
Dichte: 1.64 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Identification Passes Test
USP - Loss on Drying at 105°C ≤0.2 %
USP - Residue on Ignition ≤0.1 %
USP - Chloride (Cl) ≤0.007 %
USP - Sulfate (SO₄) ≤0.0065 %
USP - Hydrolyzable Substances Passes Test
USP - Assay (C₂H₅NO₂) (dried basis) 98.5 - 101.5 %
USP - Endotoxin Concentration (EU/g) ≤2.5
USP - Related Compounds: Glycine anhydride ≤0.1 %
USP - Related Compounds: Monochloroacetic acid (For Information Only) Passes Test
USP - Related Compounds: Iminodiacetic acid ≤0.1 %
USP - Related Compounds: Diglycine ≤0.1 %
USP - Related Compounds: Triglycine ≤0.1 %
USP - Related Compounds: Hexamethylenetetramine ≤0.1 %
USP - Related Compounds: Any unspecified impurity (For Information Only) = 0.10 %
USP - Related Compounds: Total impurities ≤1.0 %
EP/BP - Assay (C₂H₅NO₂) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 5.9 - 6.4
EP/BP - Chloride (Cl) ≤75 ppm
EP/BP - Loss on Drying at 105°C ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
EP/ BP - Related substances: Glycine anhydride ≤0.10 %
EP/ BP - Related substances: Diglycine ≤0.10 %
EP/ BP - Related substances: Triglycine ≤0.10 %
EP/ BP - Related substances: Unspecified impurities, each ≤0.10 %
EP/ BP - Related substances: Total impurities ≤0.2 %
EP/BP - Ninhydrin Positive Substances - Each Impurity ≤0.10 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤1.0 %
EP/BP - Ammonium (NH₄) ≤0.02 %
JP - Assay (C₂H₅NO₂) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - pH (1 in 20) 5.6 - 6.6
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.021 %
JP - Sulfate (SO₄) ≤0.028 %
JP - Ammonium (NH₄) ≤0.02 %
JP - Heavy Metals (as Pb) ≤20 ppm
JP - Arsenic (As) ≤2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤0.30 %
JP - Residue on Ignition ≤0.10 %
Only Class 2 Solvents (Methanol) are likely to be present. All are
below Option 1 limits.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR