di-Ammoniumhydrogencitrat, high_purity

Lieferant: VWR Chemicals
Achtung

Synonyms: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

0470-500GEA 237 CHF
0470-500G
di-Ammoniumhydrogencitrat, high_purity
di-Ammoniumhydrogencitrat
Formel: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
Dichte: 1.483 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013068
CAS-Nummer: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Order Now

Spezifikation Testergebnisse

Chloride <0.003%
Heavy Metals <0.003%
Purity ≥99.0%
Residue after ignition ≤0.05%
Sulphate ≤0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR