Natriumchlorid, crystals, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor

Synonyms: Saline, NaCl, Sea salt, Halite

0278.1000EA 45.5 CHF
0278.1000 0278.9050
Natriumchlorid, crystals, BAKER ANALYZED®, J.T.Baker®
Natriumchlorid
Formel: NaCl
MW: 58.44 g/mol
Siedepunkt: 1413
Melting Pt: 801
Dichte: 2.16 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003477
CAS-Nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Spezifikation Testergebnisse

Assay min. 99.5 %
Barium (Ba) max. 0.002 %
Bromide (Br) max. 0.01 %
Calcium (Ca) max. 0.002 %
Iodide (I) max. 0.005 %
Potassium (K) max. 0.01 %
Magnesium (Mg) max. 0.001 %
Insoluble Matter max. 0.005 %
pH of 5% Solution at 25°C 5.0 - 8.0
Sulfate (SO4) max. 0.01 %
Trace Impurities (in ppm)
Arsenic (As) max. 3
Iron (Fe) max. 3
Heavy Metals (as Pb) max. 5
Phosphate (PO4) max. 5

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR