4-(Dimethylamino)benzaldehyd, Supelco®

Lieferant: Merck

Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde

M-Clarity™ quality level = MQ100

02560-500ML
02560-500MLEA 275 CHF
02560-500ML
4-(Dimethylamino)benzaldehyd, Supelco®
4-(Dimethylamino)benzaldehyd
Formel: (CH₃)₂NC₆H₄CHO
Siedepunkt: 176…177 °C (1013 hPa)
Melting Pt: 72 °C
Dichte: 1.0254 g/cm³ (99.9 °C)
Flash-Pt: >100 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003381
CAS-Nummer: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac95b977bd89a4d2be64f1752a40d0509b4dc467&vid=e82ac1f1cec69908fe8e6c5d14e2e866eac4bb35&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR