Mangan(II)sulfat Monohydrat, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyms: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate

0174.1000EA 164 CHF
0174.1000
Mangan(II)sulfat Monohydrat, BAKER ANALYZED® ACS, J.T.Baker®
Mangan(II)sulfat Monohydrat
Formel: MnSO₄·1H₂O
Melting Pt: 700 °C
Dichte: 2.95 g/cm³ (25 °C)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00149159
CAS-Nummer: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 13,05763

Order Now

Spezifikation Testergebnisse

Exceeds ACS Specifications
Assay 98.0 - 101.0 %
Calcium (Ca) max. 0.005 %
Chloride (Cl) max. 0.001 %
Iron (Fe) max. 0.001 %
Potassium (K) max. 0.005 %
Magnesium (Mg) max. 0.005 %
Sodium (Na) max. 0.005 %
Zinc (Zn) max. 0.005 %
Heavy Metals (as Pb) max. 0.002 %
Insoluble Matter max. 0.01 %
Loss on Ignition 10.0 - 12.0 %
Substances Reducing KMnO4 passes test
Trace Impurities (in ppm)
Copper (Cu) max. 5
Nickel (Ni) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR