Phenol 99,0, Crystalline, Ultra Pure Grade

Lieferant: VWR Chemicals
Gefahr

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

0126-100GEA 169 CHF
0126-100G 0126-500G
Phenol 99,0, Crystalline, Ultra Pure Grade
Phenol
Formel: C₆H₅OH
MW: 94.11 g/mol
Siedepunkt: 181.7
Melting Pt: 40.8
Dichte: 1.06 g/cm³ (20 °C)
Flash-Pt: 81 °C (geschlossener Tiegel)
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00002143
CAS-Nummer: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
Merck Index: 12,07390

Order Now

Spezifikation Testergebnisse

Heavy Metals (as Pb) <0.01 %
Melting Point 40-41°C
Moisture (KF) <= 0.5 %
pH (Saturated) @25C Reported
Purity >=99.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR