Process chromatography resins and media
Avantor J.T.Baker® brand BAKERBOND® process chromatography products features a unique multi-mode chemistry designed to increase yield by improving the efficiency of your purification process and enhance selectivity by removing closely related impurities. Our resins and media are proven to reduce total purification steps while also improving purity and yield in bioprocessing applications.
A solution for each step in the process
The J.T.Baker® brand BAKERBOND® process chromatography offering includes a range of trusted products, suitable for the protein A capture step in mAbs manufacturing, through polishing steps in mAbs, vaccine production, recombinant protein production and more.
Expertise and support to optimize downstream production
With over four decades of experience in chromatography, our team of experts can also assist with a range of support—from column packing to custom-made media. We also offer our expertise in downstream process optimization, working with you to recommend the proper selection of buffer cleaning and solutions for your application. Learn more about our downstream process optimization services.
A legacy of excellence in the most demanding applications
For more than a century, customers around the world have trusted Avantor J.T.Baker® brand products to meet the needs of some of the most demanding applications.
Coupled with our trusted offering of cGMP chemicals and excipients, J.T.Baker® brand process chromatography media, columns and resins offer exacting performance in the critical chromatography steps in mAbs downstream processing.
Browse our range of cGMP chemicals and excipients from J.T.Baker and other brands.
Produktauswahl
J.T.Baker® BAKERBOND® PROchievA™ resin
Recombinant protein A resin offering superior dynamic binding capacity with excellent alkaline stability in mAbs manufacturing.
J.T.Baker® BAKERBOND® PROchievA™ recombinant protein A resin in pre-packaged lab columns
Suitable for use in mAbs manufacturing with a range of common chromatography instruments in convenient 1ml and 5ml lab columns.
J.T.Baker® BAKERBOND® PolyPEI bulk resin
Multimode ion exchange (IEX) chromatography resin to enhance the separation power of your chromatography process steps.
J.T.Baker® BAKERBOND® PolyPEI resin in pre-packaged lab columns
Multimode ion exchange (IEX) chromatography resin in convenient 1ml and 5ml lab columns.
J.T.Baker® BAKERBOND® PolyCSx bulk resin
Multimode ion exchange (IEX) chromatography resin with unique selectivity to enable higher purification performance.
J.T.Baker® BAKERBOND® PolyCSx resin in pre-packaged lab columns
Multimode ion exchange (IEX) chromatography resin with unique selectivity to enable higher purification performance.
J.T.Baker® BAKERBOND® Poly Hi-Propyl bulk resin
Mixed mode hydrophobic interaction chromatography (HIC) resin.
J.T.Baker® BAKERBOND® Poly Hi-Propyl resin in pre-packaged lab columns
Mixed mode hydrophobic interaction chromatography (HIC) resin in convenient 1ml and 5ml lab columns.
J.T.Baker® BAKERBOND® PolyQUAT bulk resin
Multimode ion exchange (IEX) chromatography resin to enhance the separation power of your chromatography process steps.
J.T.Baker® BAKERBOND® PolyQUAT resin in pre-packaged lab columns
Multimode ion exchange (IEX) chromatography resin in convenient 1ml and 5ml lab columns.
J.T.Baker® BAKERBOND® PolyABx bulk resin
Multimode ion exchange (IEX) chromatography resin for process chromatography.