Tridemorph

Lieferant: EHRENSTORFER GMBH

EHERC17820000EA 102 CHF
EHERC17820000
Tridemorph
Tridemorph
Formel: C₁₉H₃₉NO
CAS-Nummer: 24602-86-6
EINECS: 246-347-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR