Boc-D-Pro-OH
Lieferant: Novabiochem (Part of Merck)
04-13-0027-25EA
220
CHF
04-13-0027-25
Boc-D-Pro-OH
Boc-D-Pro-OH
Formel:
C₁₀H₁₇NO₄ MW: 215,25 g/mol |
MDL Number:
MFCD00063226 CAS-Nummer: 37784-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...